Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-phenylbutyric acid | 4593-90-2 | MFCD00002725 | 99.9% | 164.20 | C10H12O2 | 100 MG
SDP

Catalog No. 5000300346
Encompass_Preferred

3-Phenylbutyric acid (CAS 4593-90-2) is an endogenous metabolite provided as a research-grade reference standard for chemical and biological studies. It is supplied with high reported purity and storage recommendations to maintain stability; intended for research use only.

  • Endogenous metabolite useful as a biochemical reference in research.
  • Purity 99.94% as reported by the supplier.
  • Molecular formula C10H12O2; molecular weight 164.20.
  • White to light yellow; solid below 35°C and liquid above 38°C.
  • Available as solid and solution formats with multiple pack sizes.
  • Storage: -20°C for long term; shorter stability in solvent.

Catalog No. 50-003-00346 Supplier Medchemexpress LLC Supplier No. HYW017189100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Phenylbutyric acid (CAS 4593-90-2) is an endogenous metabolite provided as a research-grade reference standard for chemical and biological studies. It is supplied with high reported purity and storage recommendations to maintain stability; intended for research use only.

  • Endogenous metabolite useful as a biochemical reference in research.
  • Purity 99.94% as reported by the supplier.
  • Molecular formula C10H12O2; molecular weight 164.20.
  • White to light yellow; solid below 35°C and liquid above 38°C.
  • Available as solid and solution formats with multiple pack sizes.
  • Storage: -20°C for long term; shorter stability in solvent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.