Learn More
Medchemexpress LLC 3-phenylbutyric acid | 4593-90-2 | MFCD00002725 | 99.9% | 164.20 g/mol | C10H12O2 | 1 ML
SDP
Catalog No. 5000299893
Encompass_Preferred
3-Phenylbutyric acid is an endogenous small-molecule carboxylic acid provided as a ready-to-use 10 mM solution in DMSO (1 mL). It is intended for research use as a reagent or reference standard, offering a high-purity formulation suitable for biochemical, metabolic, and analytical workflows.
  • High purity for reliable experimental results.
  • Ready-to-use 10 mM solution in DMSO for rapid assay setup.
  • Small 1 mL volume appropriate for limited-use experiments.
  • Stable solution formulation for consistent stock concentration.
  • Applicable to biochemical, metabolic, and analytical studies.
Catalog No. 50-002-99893 Supplier Medchemexpress LLC Supplier No. HYW01718910MM/1ML
3-Phenylbutyric acid is an endogenous small-molecule carboxylic acid provided as a ready-to-use 10 mM solution in DMSO (1 mL). It is intended for research use as a reagent or reference standard, offering a high-purity formulation suitable for biochemical, metabolic, and analytical workflows.
  • High purity for reliable experimental results.
  • Ready-to-use 10 mM solution in DMSO for rapid assay setup.
  • Small 1 mL volume appropriate for limited-use experiments.
  • Stable solution formulation for consistent stock concentration.
  • Applicable to biochemical, metabolic, and analytical studies.