Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-phenylbutyric acid | 4593-90-2 | MFCD00002725 | 99.9% | 164.20 g/mol | C10H12O2 | 500 MG
SDP

Catalog No. 5000300902
Encompass_Preferred

3-Phenylbutyric acid is an endogenous metabolite reagent intended for biochemical and microbiological research. The manufacturer datasheet provides molecular and physicochemical data, solubility guidelines for in vitro and in vivo use, storage recommendations, and intended research applications.

  • Purity 99.94%.
  • Molecular weight 164.20 g/mol.
  • Chemical formula C10H12O2.
  • Soluble in DMSO at 100 mg/mL (may require ultrasonic).
  • In vivo formulations reach ≥2.5 mg/mL in common vehicles as clear solutions.
  • Storage: pure form -20 °C (up to 3 years) or 4 °C (2 years); in solvent -80 °C (6 months).

Catalog No. 50-003-00902 Supplier Medchemexpress LLC Supplier No. HYW017189500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Phenylbutyric acid is an endogenous metabolite reagent intended for biochemical and microbiological research. The manufacturer datasheet provides molecular and physicochemical data, solubility guidelines for in vitro and in vivo use, storage recommendations, and intended research applications.

  • Purity 99.94%.
  • Molecular weight 164.20 g/mol.
  • Chemical formula C10H12O2.
  • Soluble in DMSO at 100 mg/mL (may require ultrasonic).
  • In vivo formulations reach ≥2.5 mg/mL in common vehicles as clear solutions.
  • Storage: pure form -20 °C (up to 3 years) or 4 °C (2 years); in solvent -80 °C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.