Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenylglutaric acid, 98%, Thermo Scientific™

Catalog No. AC184480100
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC184480100 10 g
AC184480050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC184480100 Supplier Thermo Scientific Chemicals Supplier No. 184480100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4165-96-2
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.20
MDL Number MFCD00002718
InChI Key RZOKZOYSUCSPDF-UHFFFAOYSA-L
Synonym 3-phenylglutaric acid, pentanedioic acid, 3-phenyl, 3-phenyl-pentanedioic acid, acmc-20amyr, #
PubChem CID 77823
IUPAC Name 3-phenylpentanedioate
SMILES [O-]C(=O)CC(CC([O-])=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5CH(CH2CO2H)2
Quantity 10 g
Beilstein 09, 878
Formula Weight 208.21
Chemical Name or Material 3-Phenylglutaric acid, 98%
EINECSNumber 224-016-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.