Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Phenyllactic acid

Catalog No. P725110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P725110G 10 g
P72511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P725110G Supplier Sigma Aldrich Supplier No. P725110G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reagent involved in biological studies of:
  • Enantioselectivity of lipase in transesterification
  • Oxidation by glycolate oxidase and catalase

Specifications

CAS 828-01-3
Molecular Formula C9H10O3
Linear Formula C6H5CH2CH(OH)COOH
MDL Number MFCD00065928
Quantity 10 g
Synonym alpha-Hydroxyhydrocinnamic acid; beta-Phenyllactic acid; 2-Hydroxy-3-phenylpropionic acid
Molecular Weight (g/mol) 166.17
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.