Learn More
Cayman Chemical 3-Phenylpropanoyl-Coenzyme A
SDP
Catalog No. 502942133
Encompass_Preferred
A medium-chain and phenyl-substituted acyl CoA; has been used as an enzymatic substrate in potential screening assays for MCAD deficiency in newborn-derived primary fibroblasts
Catalog No. 50-294-2133 Supplier Cayman Chemical Supplier No. 39998500 UG
A medium-chain and phenyl-substituted acyl CoA; has been used as an enzymatic substrate in potential screening assays for MCAD deficiency in newborn-derived primary fibroblasts