Learn More
Medchemexpress LLC 3-Phosphoglyceric acid | 820-11-1 | 99.9% | 186.06 | 10 MG
SDP
Catalog No. 5000437948
Encompass_Preferred
3-Phosphoglyceric acid is a metabolic intermediate crucial for both glycolysis and the Calvin cycle, playing a role in fundamental cellular energy production and carbon fixation pathways. It is also involved in the epigenetic regulation of alveolar macrophages, influencing gene expression and cellular function in the immune system.
  • Enhances Spp1 transcription in VHL-deficient macrophages.
  • Reverses reduced H3K4me3 modification in VHL-deficient macrophages.
  • Augments gene expression and H3K4me3 deposition of Spp1 in IL-4-stimulated macrophages.
  • Acts as a metabolic checkpoint in yeast, regulating H3K4 methylation and histone phosphorylation.
Catalog No. 50-004-37948 Supplier Medchemexpress LLC Supplier No. HY11349110MG
3-Phosphoglyceric acid is a metabolic intermediate crucial for both glycolysis and the Calvin cycle, playing a role in fundamental cellular energy production and carbon fixation pathways. It is also involved in the epigenetic regulation of alveolar macrophages, influencing gene expression and cellular function in the immune system.
  • Enhances Spp1 transcription in VHL-deficient macrophages.
  • Reverses reduced H3K4me3 modification in VHL-deficient macrophages.
  • Augments gene expression and H3K4me3 deposition of Spp1 in IL-4-stimulated macrophages.
  • Acts as a metabolic checkpoint in yeast, regulating H3K4 methylation and histone phosphorylation.