Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Phosphoglyceric acid | 820-11-1 | 99.9% | 186.06 | 50 MG
SDP

Catalog No. 5000435592
Encompass_Preferred

3-Phosphoglyceric acid is a metabolic intermediate found in both glycolysis and the Calvin cycle. It plays a role in alveolar macrophage epigenetic regulation. This product is for research use only and not intended for patient use.

  • Metabolic intermediate: involved in glycolysis and the Calvin cycle.
  • Epigenetic regulation: participates in alveolar macrophage epigenetic regulation.
  • Purity: 99.88%.
  • Appearance: viscous liquid, colorless to light yellow.
  • Storage: pure form: -20°C for 3 years.
  • Storage: in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-004-35592 Supplier Medchemexpress LLC Supplier No. HY11349150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Phosphoglyceric acid is a metabolic intermediate found in both glycolysis and the Calvin cycle. It plays a role in alveolar macrophage epigenetic regulation. This product is for research use only and not intended for patient use.

  • Metabolic intermediate: involved in glycolysis and the Calvin cycle.
  • Epigenetic regulation: participates in alveolar macrophage epigenetic regulation.
  • Purity: 99.88%.
  • Appearance: viscous liquid, colorless to light yellow.
  • Storage: pure form: -20°C for 3 years.
  • Storage: in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.