Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-phosphoglyceric acid | 820-11-1 | MFCD20638229 | 99.9% | 186.06 g·mol⁻¹ | C3H7O7P | 5 MG
SDP

Catalog No. 5000263946
Encompass_Preferred

3-phosphoglyceric acid is a metabolic intermediate in glycolysis and the Calvin cycle, supplied as a high-purity biochemical reagent for research studies including metabolism and epigenetic regulation.

  • High purity (99.88%).
  • Molecular weight 186.06 g·mol⁻¹.
  • Molecular formula C3H7O7P.
  • Available in small research pack sizes such as 5 mg.
  • Suitable for studies of glycolysis, carbon fixation, and macrophage epigenetic regulation.
  • Supplied as a solid for laboratory handling and storage.

Catalog No. 50-002-63946 Supplier Medchemexpress LLC Supplier No. HY1134915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-phosphoglyceric acid is a metabolic intermediate in glycolysis and the Calvin cycle, supplied as a high-purity biochemical reagent for research studies including metabolism and epigenetic regulation.

  • High purity (99.88%).
  • Molecular weight 186.06 g·mol⁻¹.
  • Molecular formula C3H7O7P.
  • Available in small research pack sizes such as 5 mg.
  • Suitable for studies of glycolysis, carbon fixation, and macrophage epigenetic regulation.
  • Supplied as a solid for laboratory handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.