Learn More
Electron Microscopy Sciences SEM Sample Holder 3 Pin Holder, With Pin B, 25 mm dia x 10 mm H
SDP
Catalog No. 5028678
Encompass_Preferred
Our scanning electron microscope specimen holders are designed to improve your productivity and allow you to view more than one sample at a time. You will save pump down time, keep your chamber cleaner and get more work done. All mounts are machined from solid aluminum and come with spring clips/or setscrews to hold your specimens securely. All mounts are made to fit onto your stage and are designed to fit through all standard specimen exchange ports, and have a center-threaded port to accept the Adapter Pins.
All adapters have a threaded portion 5 mm in length. For 1/2" dia. (12.5mm) surface, 1/8" dia. (3.1 mm) pins. Accommodates 3. Pin overall measures: 28mm long x 6mm diameter
Catalog No. 50-286-78 Supplier Electron Microscopy Sciences Supplier No. 7592002
Our scanning electron microscope specimen holders are designed to improve your productivity and allow you to view more than one sample at a time. You will save pump down time, keep your chamber cleaner and get more work done. All mounts are machined from solid aluminum and come with spring clips/or setscrews to hold your specimens securely. All mounts are made to fit onto your stage and are designed to fit through all standard specimen exchange ports, and have a center-threaded port to accept the Adapter Pins.
All adapters have a threaded portion 5 mm in length. For 1/2" dia. (12.5mm) surface, 1/8" dia. (3.1 mm) pins. Accommodates 3. Pin overall measures: 28mm long x 6mm diameter