Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Piperidin-1-ylmethyl)benzoic acid hydrochloride hemihydrate, ≥97%, Thermo Scientific™

Catalog No. CC45251CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC45251CB 250 mg Amber Glass Bottle
CC45251DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC45251CB Supplier Thermo Scientific Chemicals Supplier No. CC45251CB
Add to Cart

Chemical Identifiers

CAS 863991-96-2
Molecular Formula C26H38Cl2N2O5
Molecular Weight (g/mol) 529.50
MDL Number MFCD07772860
InChI Key HTWIFQGKFFGIFX-UHFFFAOYSA-N
Synonym 3-piperdin-1-ylmethyl benzoic acid hydrochloride hemihydrate, bis 3-piperidin-1-ylmethyl benzoic acid hydrate dihydrochloride, benzoic acid,3-1-piperidinylmethyl-,hydrochloride,hydrate 2:2:1, 3-piperidin-1-yl methyl benzoic acid-hydrogen chloride-water 2/2/1
PubChem CID 44119333
IUPAC Name 3-(piperidin-1-ylmethyl)benzoic acid;hydrate;dihydrochloride
SMILES O.Cl.Cl.OC(=O)C1=CC(CN2CCCCC2)=CC=C1.OC(=O)C1=CC(CN2CCCCC2)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 264.76
Percent Purity 97%
Chemical Name or Material 3-(Piperidinomethyl)benzoic acid hydrochloride 0.5 hydrate
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.