Learn More
Medchemexpress LLC 3-piperidinecarboxamide, 1-[3,5-bis(trifluoromethyl)benzoyl]-N-(4-chlorophenyl) | 1207113-88-9 | 99.3% | 100 MG

Supplier: Medchemexpress LLC HY120855100MG
CCG-100602 is a small-molecule inhibitor that selectively blocks myocardin-related transcription factor A (MRTF-A) / serum response factor (SRF) signaling by preventing MRTF-A nuclear localization; supplied as a white to off-white solid for research use.
- Selective MRTF-A/SRF pathway inhibitor.
- CAS 1207113-88-9.
- Molecular weight 478.82 g/mol.
- Purity 99.29% (LCMS).
- Available as solid (5-500 mg) and as 10 mM DMSO solutions.
- Store solid at 4°C; in solvent store at -80°C for long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.