Learn More
Medchemexpress LLC 3-piperidinecarboxamide, 1-(cyclopropylmethyl)-4-[[[5-(2,4-difluorophenyl)-3-isoxazolyl]carbonyl]am | 2178049-58-4 | 98.8% | 522.55 | C27H28F2N6O3 | 5 MG
SDP
Catalog No. 5000221277
Encompass_Preferred
ACT-1004-1239 is a potent, selective, orally active CXCR7 (ACKR3) antagonist supplied for research use. It shows nanomolar potency in biochemical assays and has demonstrated target engagement and pharmacological effects in preclinical models, including modulation of CXCL12/CXCL11 signaling and efficacy in rodent disease models. Supplied formats support both in vitro and in vivo study designs.
  • Potent CXCR7 antagonism with reported IC50 of 3.2 nM.
  • High purity suitable for research applications.
  • Available as solid and 10 mM DMSO solution for convenient use.
  • Appropriate for in vitro and in vivo pharmacology studies.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-21277 Supplier Medchemexpress LLC Supplier No. HY1426175MG
ACT-1004-1239 is a potent, selective, orally active CXCR7 (ACKR3) antagonist supplied for research use. It shows nanomolar potency in biochemical assays and has demonstrated target engagement and pharmacological effects in preclinical models, including modulation of CXCL12/CXCL11 signaling and efficacy in rodent disease models. Supplied formats support both in vitro and in vivo study designs.
  • Potent CXCR7 antagonism with reported IC50 of 3.2 nM.
  • High purity suitable for research applications.
  • Available as solid and 10 mM DMSO solution for convenient use.
  • Appropriate for in vitro and in vivo pharmacology studies.
  • For research use only; not for human or veterinary use.