Learn More
Medchemexpress LLC 3-Piperidinecarboxylic acid, 2-[4-(cyclopentylamino)phenyl]-1-(2-fluoro-6-methylbenzoyl)-, (2R, 3S)- | 1346623-11-7 | 98.0% | 424.51 | 500 MG
SDP
Catalog No. 5000403570
Encompass_Preferred
(2R,3S)-2-(4-(Cyclopentylamino)phenyl)-1-(2-fluoro-6-methylbenzoyl)piperidine-3-carboxylic acid is a drug intermediate for the synthesis of various active compounds.
  • Suitable for synthesis of active compounds.
  • Multiple storage options for powder and in solvent.
  • Available as a light yellow to brown solid.
  • 1H NMR spectrum is consistent with structure.
Catalog No. 50-004-03570 Supplier Medchemexpress LLC Supplier No. HY42565500MG
(2R,3S)-2-(4-(Cyclopentylamino)phenyl)-1-(2-fluoro-6-methylbenzoyl)piperidine-3-carboxylic acid is a drug intermediate for the synthesis of various active compounds.
  • Suitable for synthesis of active compounds.
  • Multiple storage options for powder and in solvent.
  • Available as a light yellow to brown solid.
  • 1H NMR spectrum is consistent with structure.