Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Piperidino-1,2-propanediol 96%, Thermo Scientific™

Catalog No. AC203841000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC203841000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC203841000 Supplier Thermo Scientific Chemicals Supplier No. 203841000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4847-93-2
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.229
InChI Key MECNWXGGNCJFQJ-UHFFFAOYSA-N
Synonym 3-piperidino-1,2-propanediol, 3-piperidin-1-yl propane-1,2-diol, 1-n-piperidino-2,3-propanediol, 3-piperidinopropane-1,2-diol, 3-piperidylpropane-1,2-diol, 1-piperidino-2,3-propanediol, 1,2-propanediol,3-1-piperidinyl, 1,2-propanediol, 3-1-piperidinyl, 3-1-piperidinyl-1,2-propanediol #
PubChem CID 97824
IUPAC Name 3-piperidin-1-ylpropane-1,2-diol
SMILES C1CCN(CC1)CC(CO)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Quantity 100 g
Beilstein 20, 34
Formula Weight 159.23
Chemical Name or Material 3-Piperidino-1, 2-propanediol, 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.