Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Piperidinobenzaldehyde, 95%, Thermo Scientific™

Catalog No. CC34004CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC34004CB 250 mg Amber Glass Bottle
CC34004DA 1 g Amber Glass Bottle
CC34004DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC34004CB Supplier Thermo Scientific Chemicals Supplier No. CC34004CB
Add to Cart

Chemical Identifiers

CAS 669050-72-0
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
InChI Key FXXQUTSXXILOMY-UHFFFAOYSA-N
Synonym 3-piperidinobenzaldehyde, 3-piperidin-1-yl benzaldehyde, benzaldehyde, 3-1-piperidinyl, 3-piperidylbenzaldehyde, 3-1-piperidinyl benzaldehyde
PubChem CID 7164587
IUPAC Name 3-piperidin-1-ylbenzaldehyde
SMILES C1CCN(CC1)C2=CC=CC(=C2)C=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 189.26
Percent Purity 95%
Chemical Name or Material 3-Piperidinobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.