Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[3-(Piperidinomethyl)phenyl]methanol, ≥90%, Thermo Scientific™

Catalog No. CC45209DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC45209DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC45209DA Supplier Thermo Scientific Chemicals Supplier No. CC45209DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73278-91-8
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.301
MDL Number MFCD07772858
InChI Key NZKXLHGCZWDXGE-UHFFFAOYSA-N
Synonym 3-piperidinomethyl phenyl methanol, 3-piperidin-1-ylmethyl phenyl methanol, 3-piperidinomethylbenzylalcohol, 3-1-piperidylmethyl-benzyl alcohol, 3-1-piperidinylmethyl benzenemethanol, 3-piperidylmethyl phenyl methan-1-ol
PubChem CID 7164645
IUPAC Name [3-(piperidin-1-ylmethyl)phenyl]methanol
SMILES C1CCN(CC1)CC2=CC(=CC=C2)CO

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.3
Percent Purity 90%
Chemical Name or Material [3-(Piperidinomethyl)phenyl]methanol
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.