Learn More
3-Piperidylpropiophenone Hydrochloride, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 886-06-6 |
| Physical Form | Solid |
| Quantity | 50 mg |
| Molecular Formula | C14H19NO · HCl |
| Linear Formula | C14H19NO · HCl |
| Synonym | 1-Phenyl-3-(piperidin-1-yl)propan-1-one hydrochloride; 3-Piperidinopropiophenone hydrochloride |
| Molecular Weight (g/mol) | 253.77 |
| Formula Weight | 253.77 |
| Grade | BP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.