Learn More
Research Products International Corp 3 Inch Platform Head with Rubber Cover for Vortex Genie Mixers
SDP
Catalog No. 50998321
Encompass
Plastic 3-inch (76mm) platform with rubber cover for mixing and vortexing in beakers, flasks and multiple tubes. The plastic 3-inch platform with rubber cover may be used on the Vortex-Genie 1, Vortex-Genie Pulse and the Vortex-Genie 2 family of mixers.
Catalog No. 50-998-321 Supplier Research Products International Corp Supplier No. 155312
Plastic 3-inch (76mm) platform with rubber cover for mixing and vortexing in beakers, flasks and multiple tubes. The plastic 3-inch platform with rubber cover may be used on the Vortex-Genie 1, Vortex-Genie Pulse and the Vortex-Genie 2 family of mixers.