Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100D RX100024
Encompass_Preferred
3 place multi position combo mantle for 1L round bottom flask, to 400 degrees C internal operating temperature. 290W, 240V
Catalog No. NC1843348