Learn More
Ace Glass, Inc. Complete vacuum glass manifold unit with three 4mmstraight bore PTFE stopcocks. Unit includes a #15 end plug and two Ace-Safe tubing connectors for 1/4" and 1/2" tubing.
SDP
Supplier:  Ace Glass, Inc. 876344
Encompass
Complete vacuum glass manifold unit with three 4mmstraight bore PTFE stopcocks. Unit includes a #15 end plug and two Ace-Safe tubing connectors for 1/4" and 1/2" tubing.
Catalog No. 50-976-388