Learn More
Chemglass Life Sciences 3-Port Airfree Schlenk Inert Gas Vacuum Manifold, Double Bank, 410mm OAL
SDP
Catalog No. AF006001
Encompass_Preferred
3-Port Airfree Schlenk Inert Gas Vacuum Manifold, Double Bank, 410mm OAL. Double bank manifold available in 3, 4 or 5 port configuration. Forward facing 0-8mm Chem-Vac™ Chem-Cap valves permit easy operation of manifold inside of a fume hood. Main ports are spaced 100mm center to center with an additional 2 ports on the upper bank that can be used for gauging or sealed with AF-0210 stoppers. All connection ports are #15 o-ring joint connections that incorporate a greaseless o-ring seal. Manifold is modular in design allowing the user tremendous flexibility in performing experiments. Adapters from #15 o-ring joints to standard taper components, serrated hose connections or 3/8" Cajon fittings are available to allow the ports to be used with existing glassware. Replacement Chem-Vac™ Chem-Cap plug is listed below.
Catalog No. AF006001 Supplier Chemglass Life Sciences Supplier No. AF006001
3-Port Airfree Schlenk Inert Gas Vacuum Manifold, Double Bank, 410mm OAL. Double bank manifold available in 3, 4 or 5 port configuration. Forward facing 0-8mm Chem-Vac™ Chem-Cap valves permit easy operation of manifold inside of a fume hood. Main ports are spaced 100mm center to center with an additional 2 ports on the upper bank that can be used for gauging or sealed with AF-0210 stoppers. All connection ports are #15 o-ring joint connections that incorporate a greaseless o-ring seal. Manifold is modular in design allowing the user tremendous flexibility in performing experiments. Adapters from #15 o-ring joints to standard taper components, serrated hose connections or 3/8" Cajon fittings are available to allow the ports to be used with existing glassware. Replacement Chem-Vac™ Chem-Cap plug is listed below.