Learn More
Bellco Glass, Inc. 3-Port Sparger Assembly W/Frit,15L For Vertical Side Arm Vessels
SDP
Catalog No. 501944842
Encompass_Preferred
316-l stainless steel ra32 or better finish10 micron porosity stainless steel frit for low profile vertical side arm vessels. MADE IN THE USA. Frit is removable for cleaning. Comes with 45mm blue polypropylene open top cap and silicone gasketTriple tube configuration With Vent, Fill/Sample And Sparge.
Catalog No. 50-194-4842 Supplier Bellco Glass, Inc. Supplier No. 196461015
316-l stainless steel ra32 or better finish10 micron porosity stainless steel frit for low profile vertical side arm vessels. MADE IN THE USA. Frit is removable for cleaning. Comes with 45mm blue polypropylene open top cap and silicone gasketTriple tube configuration With Vent, Fill/Sample And Sparge.