Learn More
Bellco Glass, Inc. 3-Port Spargr Assembly W/Holes,15L For Vertical Side Arm Vessels
SDP
Catalog No. 501944847
Encompass_Preferred
316-l stainless steel ra32 or better finish.040" drilled holes sparger for low profile vertical side arm vessels. MADE IN THE USA. Threaded end is removable for cleaning.Comes with 45mm blue polypropylene open top cap and silicone gasket.Triple tube configuration With Vent, Fill/Sample And Sparge.
Catalog No. 50-194-4847 Supplier Bellco Glass, Inc. Supplier No. 196462015
316-l stainless steel ra32 or better finish.040" drilled holes sparger for low profile vertical side arm vessels. MADE IN THE USA. Threaded end is removable for cleaning.Comes with 45mm blue polypropylene open top cap and silicone gasket.Triple tube configuration With Vent, Fill/Sample And Sparge.