Learn More
Bellco Glass, Inc. 3-Port Spargr Assembly W/Holes,36L For Vertical Side Arm Vessels
SDP
Catalog No. 501944848
Encompass_Preferred
MADE IN THE USA. Material: 316L SST with Ra Finish 32µ or better - inside & outTubing Specifications: .250" OD x .152" IDVent Port: 1.0" long - above & below cap slugAddition Port: 1.0" long above cap slug & 16 1/14"(approx) below cap slugMedia Port: 1.0" long above cap slug with 1 3/4" bend radius & 12 25/32" longTen (10) drilled holes (.040"). Supply Cap & Liner
Catalog No. 50-194-4848 Supplier Bellco Glass, Inc. Supplier No. 196462036
MADE IN THE USA. Material: 316L SST with Ra Finish 32µ or better - inside & outTubing Specifications: .250" OD x .152" IDVent Port: 1.0" long - above & below cap slugAddition Port: 1.0" long above cap slug & 16 1/14"(approx) below cap slugMedia Port: 1.0" long above cap slug with 1 3/4" bend radius & 12 25/32" longTen (10) drilled holes (.040"). Supply Cap & Liner