Learn More
Chemglass Life Sciences Addition Tube Assembly, 3-Port, SST, with Hose Barbs, for VSA Vessels
SDP
Catalog No. 50143200
Encompass
Our hose barbed 3-port dip tube assemblies are designed for addition or sampling of media from Vertical Side Arm (VSA) Bioreactor Vessels.These addition dip tube assemblies are ideal for liquid addition of acid, base, antifoam or media.They are constructed of 316L stainless steel, electropolished to a 20Ra (or better) finish and include a blue, polypropylene GL45 open top cap and white silicone gasket.Tube lengths are 3-3/32” long.
Catalog No. 50-143-200 Supplier Chemglass Life Sciences Supplier No. CLS1437A10
Our hose barbed 3-port dip tube assemblies are designed for addition or sampling of media from Vertical Side Arm (VSA) Bioreactor Vessels.These addition dip tube assemblies are ideal for liquid addition of acid, base, antifoam or media.They are constructed of 316L stainless steel, electropolished to a 20Ra (or better) finish and include a blue, polypropylene GL45 open top cap and white silicone gasket.Tube lengths are 3-3/32” long.