Learn More
Bellco Glass, Inc. 3 Port Stainless Steel Assembly ,36L For Vertical Over Head Drive Vessels
SDP
Catalog No. 501944789
Encompass_Preferred
Ideal for use with over head drive vessels. Manufactured from 316l electropolished stainless steel with 32ra finish or better. MADE IN THE USA. 3-PORT CAP ASSY FOR 36L OHD VESSEL. WITH 3 SST 1/4" OD TUBES. Longest tube over 18". MEDIUM TUBE:3 3/16". VENT TUBE: 2 5/32". ALL TUBES EXTENDED 1.0" ABOVE THE CAP SLUG. SUPPLY WITH A523-599 (S/CAP) & A523-600 (LINER)
Catalog No. 50-194-4789 Supplier Bellco Glass, Inc. Supplier No. 196433600R
Ideal for use with over head drive vessels. Manufactured from 316l electropolished stainless steel with 32ra finish or better. MADE IN THE USA. 3-PORT CAP ASSY FOR 36L OHD VESSEL. WITH 3 SST 1/4" OD TUBES. Longest tube over 18". MEDIUM TUBE:3 3/16". VENT TUBE: 2 5/32". ALL TUBES EXTENDED 1.0" ABOVE THE CAP SLUG. SUPPLY WITH A523-599 (S/CAP) & A523-600 (LINER)