Learn More
Bellco Glass, Inc. 3 Port Stainless Steel Assembly , 15L For Vertical Over Head Drive Vessels
SDP
Catalog No. 501944788
Encompass_Preferred
Ideal for use with over head drive vessels. MADE IN THE USA. Manufactured from 316l electropolished stainless steel with 32ra finish or better. MADE IN THE USA. 1-siphon tube 15 5/8" long 1-fill tube 2 17/64" long 1-vent tube 1" long. 45mm 3-PORT ASSY FOR 1964 - STYLE OHD VESSEL, 8 & 15L. SUPPLY 45mM S/CAP (A523-599) & WHITE SILICONE LINER (A523-600). LENGTHS ARE MEASURED FROM TOP OF S/A TO BTTM OF TUBE
Catalog No. 50-194-4788 Supplier Bellco Glass, Inc. Supplier No. 196431015R
Ideal for use with over head drive vessels. MADE IN THE USA. Manufactured from 316l electropolished stainless steel with 32ra finish or better. MADE IN THE USA. 1-siphon tube 15 5/8" long 1-fill tube 2 17/64" long 1-vent tube 1" long. 45mm 3-PORT ASSY FOR 1964 - STYLE OHD VESSEL, 8 & 15L. SUPPLY 45mM S/CAP (A523-599) & WHITE SILICONE LINER (A523-600). LENGTHS ARE MEASURED FROM TOP OF S/A TO BTTM OF TUBE