Learn More
Bellco Glass, Inc. 3 Port Stainless Steel Assembly , 8L For Vertical Over Head Drive Vessels
SDP
Catalog No. 501944787
Encompass_Preferred
45mm 3-port assembly for 1964 style over head drive vessels, 36l. MADE IN THE USA Sealed weld – electropolish finish Supplied with 45mm s/cap & white silicone liner316l SST material, ra 32 finish or better 1 – siphon tube; 18" long 1 – fill tube; 3 3/16" 1 - VENT TUBE; 1" LONG. LENGTHS ARE MEASURED FROM TOP OF S/A TO BTTM OF TUBE
Catalog No. 50-194-4787 Supplier Bellco Glass, Inc. Supplier No. 196431008R
45mm 3-port assembly for 1964 style over head drive vessels, 36l. MADE IN THE USA Sealed weld – electropolish finish Supplied with 45mm s/cap & white silicone liner316l SST material, ra 32 finish or better 1 – siphon tube; 18" long 1 – fill tube; 3 3/16" 1 - VENT TUBE; 1" LONG. LENGTHS ARE MEASURED FROM TOP OF S/A TO BTTM OF TUBE