Learn More
Chemglass Life Sciences 3-Port SST Dip Tube Assy for 1L Spinner Flasks
SDP
Catalog No. 50143196
Encompass
Our hose barbed 3-port drip tube assemblies are designed for addition or sampling gof media from Angled Side Arm (ASA) Spinner Flasks.The dip tube assemblies are constructed of 316L stainless steel, electropolished to a 20Ra (or better) finish and include a blue PP GL-45 open top cap with white silicone gasket.
Catalog No. 50-143-196 Supplier Chemglass Life Sciences Supplier No. CLS142781
Our hose barbed 3-port drip tube assemblies are designed for addition or sampling gof media from Angled Side Arm (ASA) Spinner Flasks.The dip tube assemblies are constructed of 316L stainless steel, electropolished to a 20Ra (or better) finish and include a blue PP GL-45 open top cap with white silicone gasket.