Learn More
Chemglass Life Sciences 3-Port SST Thermowell Assy for 1L Spinner Flasks
SDP
Catalog No. 50143218
Encompass
Our hose barbed 3-port large diameter thermowell assemblies are designed to accommodate a wide variety of RTD's and dial thermometers of different diameters for measuring the process temperature inAngled Side Arm (ASA) Spinner Flasks.All 316L stainless steel tubes have an electropolished finish to a Ra 20u (or better).These assemblies feature a thermowell tube with 0. 375" OD and a 0. 305" ID that will accept all 1/4" thermometer probes.The assemblies include a blue PP open top GL-45 cap with white silicone gasket.
Catalog No. 50-143-218 Supplier Chemglass Life Sciences Supplier No. CLS142790
Our hose barbed 3-port large diameter thermowell assemblies are designed to accommodate a wide variety of RTD's and dial thermometers of different diameters for measuring the process temperature inAngled Side Arm (ASA) Spinner Flasks.All 316L stainless steel tubes have an electropolished finish to a Ra 20u (or better).These assemblies feature a thermowell tube with 0. 375" OD and a 0. 305" ID that will accept all 1/4" thermometer probes.The assemblies include a blue PP open top GL-45 cap with white silicone gasket.