Learn More
Bellco Glass, Inc. 3-Port Thermowell Assembly ,8L For Vertical Side Arm Vessels
SDP
Catalog No. 501944856
Encompass_Preferred
Thermowell assemblies are used to hold inside one tube the rtd temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. For OHD 8L Vessel. MADE IN THE USA. Material: 316L SST with Ra 32 or better - inside & outOne Vent port; .250"OD x .152" ID x 1.0"above & below cap slugOne Addition port: .250"OD x .152"ID x 15 1/8" insertion lengthOne Thermowell port: .375"OD x .305"ID x 15 1/8" insertion lengthAll tubes seam weld in to cap slug - bottom side onlySupply with 45mm PP blue open top screw cap & white silicone gasket
Catalog No. 50-194-4856 Supplier Bellco Glass, Inc. Supplier No. 196464008
Thermowell assemblies are used to hold inside one tube the rtd temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. For OHD 8L Vessel. MADE IN THE USA. Material: 316L SST with Ra 32 or better - inside & outOne Vent port; .250"OD x .152" ID x 1.0"above & below cap slugOne Addition port: .250"OD x .152"ID x 15 1/8" insertion lengthOne Thermowell port: .375"OD x .305"ID x 15 1/8" insertion lengthAll tubes seam weld in to cap slug - bottom side onlySupply with 45mm PP blue open top screw cap & white silicone gasket