Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100D RJ100024
Encompass_Preferred
Multi positon combo mantle, 3 place combo for round bottom flask, to 600 degrees C maximum internal operating temperature. 450W, 240V
Catalog No. NC1843340