Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 105UM3
Catalog No. 50871404
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 150; Sieve opening; 105um
Catalog No. 50-871-404 Supplier Gilson Company Inc Supplier No. GAA64
Add to Cart
Add to Cart
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 150; Sieve opening; 105um