Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 10UM3 P
Catalog No. 50871429
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 1000; Sieve opening; 10um
Catalog No. 50-871-429 Supplier Gilson Company Inc Supplier No. GAA83
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 1000; Sieve opening; 10um