Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 125UM3
Catalog No. 50871402
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 120; Sieve opening; 150um
Catalog No. 50-871-402 Supplier Gilson Company Inc Supplier No. GAA63
Add to Cart
Add to Cart
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 120; Sieve opening; 150um