Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 20UM3 P
Catalog No. 50871427
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 750; Sieve opening; 20um
Catalog No. 50-871-427 Supplier Gilson Company Inc Supplier No. GAA81
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 750; Sieve opening; 20um