Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 28UM3 P
Catalog No. 50871425
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 500; Sieve opening; 28um
Catalog No. 50-871-425 Supplier Gilson Company Inc Supplier No. GAA79A
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 500; Sieve opening; 28um