Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 3UM3 PR
Supplier:  Gilson Company Inc GAA84A
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 1500; Sieve opening; 3um
Catalog No. 50-871-432