Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 50UM3 P
Catalog No. 50871417
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 333; Sieve opening; 50um
Catalog No. 50-871-417 Supplier Gilson Company Inc Supplier No. GAA75
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 333; Sieve opening; 50um