Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 53UM3 P
Catalog No. 50871416
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 250; Sieve opening; 53um
Catalog No. 50-871-416 Supplier Gilson Company Inc Supplier No. GAA74A
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 250; Sieve opening; 53um