Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 55UM3 P
Catalog No. 50871415
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 250; Sieve opening; 55um
Catalog No. 50-871-415 Supplier Gilson Company Inc Supplier No. GAA74
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 250; Sieve opening; 55um