Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 5UM3 PR
Catalog No. 50871431
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 1500; Sieve opening; 5um
Catalog No. 50-871-431 Supplier Gilson Company Inc Supplier No. GAA84
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 1500; Sieve opening; 5um