Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 60UM3 P
Catalog No. 50871414
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 250; Sieve opening; 60um
Catalog No. 50-871-414 Supplier Gilson Company Inc Supplier No. GAA73
Add to Cart
Add to Cart
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 250; Sieve opening; 60um