Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 63UM3 P
Catalog No. 50871413
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 250; Sieve opening; 63um
Catalog No. 50-871-413 Supplier Gilson Company Inc Supplier No. GAA72A
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 250; Sieve opening; 63um