Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 65UM3 P
Catalog No. 50871412
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 250; Sieve opening; 65um
Catalog No. 50-871-412 Supplier Gilson Company Inc Supplier No. GAA72
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 250; Sieve opening; 65um