Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 75UM3 P
Catalog No. 50871410
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 200; Sieve opening; 75um
Catalog No. 50-871-410 Supplier Gilson Company Inc Supplier No. GAA70
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 200; Sieve opening; 75um