Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 7UM3 PR
Catalog No. 50871430
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 1000; Sieve opening; 7um
Catalog No. 50-871-430 Supplier Gilson Company Inc Supplier No. GAA83A
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 1000; Sieve opening; 7um