Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 85UM3 P
Catalog No. 50871408
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 200; Sieve opening; 85um
Catalog No. 50-871-408 Supplier Gilson Company Inc Supplier No. GAA68
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 200; Sieve opening; 85um