Learn More
Gilson Company Inc 3 PREC ACRYLIC SIEVE 90UM3 P
Catalog No. 50871407
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 200; Sieve opening; 90um
Catalog No. 50-871-407 Supplier Gilson Company Inc Supplier No. GAA67
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with unsupported electroformed nickel mesh; 3 in. dia.; Lines per inch: 200; Sieve opening; 90um